Assay Details
Gene Symbol: BAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577426.2 | XP_005577483.1 | 2 - 3 | 292 | 76 | |
XM_005577427.2 | XP_005577484.1 | 2 - 3 | 292 | 76 | ||
GenBank mRNA | AB220436.1 | - | 1 - 2 | 37 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
