Assay Details
Gene Symbol: DUSP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566410.2 | XP_005566467.1 | 2 - 3 | 585 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
