Assay Details
Gene Symbol: HAGHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015442650.1 | XP_015298136.1 | 7 - 8 | 981 | 133 | |
XM_015442651.1 | XP_015298137.1 | 7 - 8 | 982 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
