Assay Details
Gene Symbol: APOPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005562291.2 | XP_005562348.1 | 3 - 4 | 637 | 129 | |
XM_005562292.2 | XP_005562349.1 | 3 - 4 | 436 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
