Assay Details
| Gene Symbol: MAP3K21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539759.2 | XP_005539816.1 | 6 - 7 | 1946 | 109 | |
| XM_015440272.1 | XP_015295758.1 | 6 - 7 | 2000 | 109 | ||
More Information
Set Membership: |
|

MAP3K21
Genomic Map