Assay Details
Gene Symbol: LGALS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577520.2 | XP_005577577.1 | 4 - 5 | 882 | 139 | |
XM_005577522.2 | XP_005577579.1 | 3 - 4 | 758 | 139 | ||
XM_005577523.2 | XP_005577580.1 | 3 - 4 | 413 | 139 | ||
XM_015434584.1 | XP_015290070.1 | 4 - 5 | 875 | 139 | ||
XM_015434585.1 | XP_015290071.1 | 4 - 5 | 875 | 139 | ||
XM_015434586.1 | XP_015290072.1 | 4 - 5 | 875 | 139 | ||
XM_015434587.1 | XP_015290073.1 | 4 - 5 | 875 | 139 | ||
XM_015434588.1 | XP_015290074.1 | 4 - 5 | 882 | 139 | ||
XM_015434589.1 | XP_015290075.1 | 4 - 5 | 875 | 139 | ||
XM_015434590.1 | XP_015290076.1 | 4 - 5 | 875 | 139 | ||
XM_015434591.1 | XP_015290077.1 | 4 - 5 | 875 | 139 | ||
XM_015434593.1 | XP_015290079.1 | 5 - 6 | 534 | 139 | ||
XM_015434594.1 | XP_015290080.1 | 5 - 6 | 815 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
