Assay Details
| Gene Symbol: PDGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579478.2 | XP_005579535.1 | 5 - 6 | 1237 | 148 | |
| XM_005579479.2 | XP_005579536.1 | 5 - 6 | 1219 | 148 | ||
More Information
Set Membership: |
|

PDGFD
Genomic Map