Assay Details
Gene Symbol: ADCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550944.2 | XP_005551001.1 | 7 - 8 | 1916 | 112 | |
XM_005550945.2 | XP_005551002.1 | 6 - 7 | 1778 | 112 | ||
XM_005550946.2 | XP_005551003.1 | 6 - 7 | 1655 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
