Assay Details
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543302.1 | XP_005543359.1 | 3 - 4 | 844 | 134 | |
XM_005543303.2 | XP_005543360.1 | 4 - 5 | 1147 | 134 | ||
XM_015435593.1 | XP_015291079.1 | 4 - 5 | 678 | 134 | ||
XM_015435595.1 | XP_015291081.1 | 4 - 5 | 682 | 134 | ||
XM_015435597.1 | XP_015291083.1 | 3 - 4 | 746 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
