Assay Details
Gene Symbol: ZFP91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577790.2 | XP_005577847.1 | 7 - 8 | 1119 | 83 | |
XM_005577791.2 | XP_005577848.1 | 7 - 8 | 1116 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
