Assay Details
Gene Symbol: SRSF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005552380.2 | XP_005552437.1 | 3 - 4 | 426 | 81 | |
XM_005552382.2 | XP_005552439.1 | 3 - 4 | 340 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
