Assay Details
| Gene Symbol: ASB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547447.2 | XP_005547504.1 | 9 - 10 | 2099 | 102 | |
| XM_015446097.1 | XP_015301583.1 | 10 - 11 | 2214 | 102 | ||
| XM_015446098.1 | XP_015301584.1 | 10 - 11 | 2168 | 102 | ||
More Information
Set Membership: |
|

ASB14
Genomic Map