Assay Details
Gene Symbol: ZNF618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015436808.1 | XP_015292294.1 | 3 - 4 | 582 | 88 | |
XM_015436809.1 | XP_015292295.1 | 3 - 4 | 582 | 88 | ||
XM_015436810.1 | XP_015292296.1 | 3 - 4 | 582 | 88 | ||
XM_015436811.1 | XP_015292297.1 | 3 - 4 | 581 | 88 | ||
XM_015436812.1 | XP_015292298.1 | 3 - 4 | 581 | 88 | ||
XM_015436813.1 | XP_015292299.1 | 3 - 4 | 582 | 88 | ||
XM_015436816.1 | XP_015292302.1 | 3 - 4 | 582 | 88 | ||
XM_015436817.1 | XP_015292303.1 | 3 - 4 | 580 | 88 | ||
XM_015436818.1 | XP_015292304.1 | 3 - 4 | 580 | 88 | ||
XM_015436820.1 | XP_015292306.1 | 3 - 4 | 581 | 88 | ||
XM_015436821.1 | XP_015292307.1 | 3 - 4 | 579 | 88 | ||
XM_015436822.1 | XP_015292308.1 | 3 - 4 | 581 | 88 | ||
XM_015436823.1 | XP_015292309.1 | 3 - 4 | 580 | 88 | ||
XM_015436824.1 | XP_015292310.1 | 3 - 4 | 580 | 88 | ||
XM_015436827.1 | XP_015292313.1 | 3 - 4 | 581 | 88 | ||
XM_015436828.1 | XP_015292314.1 | 3 - 4 | 580 | 88 | ||
XM_015436831.1 | XP_015292317.1 | 3 - 4 | 580 | 88 | ||
XM_015436832.1 | XP_015292318.1 | 3 - 4 | 577 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
