Assay Details
Gene Symbol: SCRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583518.2 | XP_005583575.1 | 2 - 3 | 800 | 57 | |
XM_005583519.2 | XP_005583576.1 | 3 - 4 | 477 | 57 | ||
XM_005583520.2 | XP_005583577.1 | 3 - 4 | 481 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
