Assay Details
| Gene Symbol: MFSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564389.2 | XP_005564446.1 | 3 - 4 | 1493 | 89 | |
| XM_005564390.2 | XP_005564447.1 | 4 - 5 | 1356 | 89 | ||
More Information
Set Membership: |
|

MFSD3
Genomic Map