Assay Details
| Gene Symbol: ADAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592667.2 | XP_005592724.1 | 5 - 6 | 966 | 105 | |
| XM_015442713.1 | XP_015298199.1 | 6 - 7 | 1062 | 105 | ||
| XM_015442714.1 | XP_015298200.1 | 6 - 7 | 1062 | 105 | ||
| XM_015442715.1 | XP_015298201.1 | 5 - 6 | 936 | 105 | ||
| GenBank mRNA | AB070097.1 | - | 6 - 7 | 863 | 105 | |
More Information
Set Membership: |
|

ADAD2
Genomic Map