Assay Details
Gene Symbol: LGI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319455.1 | NP_001306384.1 | 1 - 2 | 465 | 122 | |
GenBank mRNA | AB169921.1 | - | 1 - 2 | 465 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
