Assay Details
| Gene Symbol: PNPLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592919.2 | XP_005592976.1 | 5 - 6 | 652 | 96 | |
| XM_005592922.2 | XP_005592979.1 | 5 - 6 | 652 | 96 | ||
| XM_015443345.1 | XP_015298831.1 | 5 - 6 | 652 | 96 | ||
| XM_015443346.1 | XP_015298832.1 | 5 - 6 | 814 | 96 | ||
| XM_015443347.1 | XP_015298833.1 | 5 - 6 | 1326 | 96 | ||
| XM_015443348.1 | XP_015298834.1 | 5 - 6 | 652 | 96 | ||
| XM_015443349.1 | XP_015298835.1 | 5 - 6 | 652 | 96 | ||
| XM_015443350.1 | XP_015298836.1 | 5 - 6 | 652 | 96 | ||
| XM_015443351.1 | XP_015298837.1 | 5 - 6 | 652 | 96 | ||
| XM_015443352.1 | XP_015298838.1 | 5 - 6 | 652 | 96 | ||
More Information
Set Membership: |
|

PNPLA4
Genomic Map