Assay Details
Gene Symbol: KRT75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570930.2 | XP_005570987.2 | 8 - 9 | 1682 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRT75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570930.2 | XP_005570987.2 | 8 - 9 | 1682 | 142 |
Set Membership: |
![]() ![]() ![]() ![]() |