Assay Details
Gene Symbol: CBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585175.2 | XP_005585232.1 | 4 - 5 | 301 | 77 | |
XM_015438920.1 | XP_015294406.1 | 4 - 5 | 291 | 77 | ||
XM_015438921.1 | XP_015294407.1 | 2 - 3 | 331 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015438920.1, XM_015438921.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
