Assay Details
| Gene Symbol: KLHL41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573404.2 | XP_005573461.1 | 2 - 3 | 1506 | 148 | |
| XM_005573405.2 | XP_005573462.1 | 2 - 3 | 1506 | 148 | ||
More Information
Set Membership: |
|

KLHL41
Genomic Map