Assay Details
Gene Symbol: EMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592708.2 | XP_005592765.1 | 1 - 2 | 477 | 121 | |
XM_005592709.2 | XP_005592766.1 | 1 - 2 | 539 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
