Assay Details
Gene Symbol: LOC102126820 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583086.2 | XP_005583143.1 | 4 - 5 | 678 | 76 | |
XM_015437751.1 | XP_015293237.1 | 4 - 5 | 684 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
