Assay Details
Gene Symbol: FOSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589591.2 | XP_005589648.1 | 2 - 3 | 1180 | 69 | |
XM_015441575.1 | XP_015297061.1 | 2 - 3 | 1063 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
