Assay Details
| Gene Symbol: ST3GAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592520.2 | XP_005592577.1 | 6 - 7 | 2096 | 115 | |
| XM_005592522.2 | XP_005592579.1 | 5 - 6 | 1923 | 115 | ||
More Information
Set Membership: |
|

ST3GAL2
Genomic Map