Assay Details
| Gene Symbol: HSPA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548936.2 | XP_005548993.1 | 3 - 4 | 784 | 138 | |
| XM_015446971.1 | XP_015302457.1 | 3 - 4 | 573 | 138 | ||
More Information
Set Membership: |
|

HSPA13
Genomic Map