Assay Details
Gene Symbol: LRRC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015430308.1 | XP_015285794.1 | 5 - 6 | 1311 | 116 | |
GenBank mRNA | AB168408.1 | - | 4 - 5 | 658 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
