Assay Details
| Gene Symbol: ZNF41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593424.2 | XP_005593481.1 | 1 - 2 | 485 | 138 | |
| XM_015443711.1 | XP_015299197.1 | 1 - 2 | 485 | 138 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005593424.2, XM_015443711.1 transcripts.
Set Membership: |
|

ZNF41
Genomic Map