Assay Details
| Gene Symbol: WBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575526.2 | XP_005575583.1 | 2 - 3 | 603 | 125 | |
| XM_015433513.1 | XP_015288999.1 | 4 - 5 | 798 | 125 | ||
| XM_015433514.1 | XP_015289000.1 | 4 - 5 | 421 | 125 | ||
| XM_015433515.1 | XP_015289001.1 | 4 - 5 | 591 | 125 | ||
| XM_015433516.1 | XP_015289002.1 | 3 - 4 | 723 | 125 | ||
| XM_015433517.1 | XP_015289003.1 | 3 - 4 | 313 | 125 | ||
| XM_015433518.1 | XP_015289004.1 | 3 - 4 | 600 | 125 | ||
| XM_015433519.1 | XP_015289005.1 | 2 - 3 | 226 | 125 | ||
More Information
Set Membership: |
|

WBP1
Genomic Map