Assay Details
Gene Symbol: CCDC106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590421.2 | XP_005590478.1 | 4 - 5 | 1713 | 70 | |
XM_005590422.2 | XP_005590479.1 | 2 - 3 | 755 | 70 | ||
XM_005590423.2 | XP_005590480.1 | 3 - 4 | 1612 | 70 | ||
GenBank mRNA | AB173691.1 | - | 3 - 4 | 1084 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
