Assay Details
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568622.2 | XP_005568679.1 | 3 - 4 | 598 | 66 | |
XM_015457882.1 | XP_015313368.1 | 3 - 4 | 790 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
