Assay Details
| Gene Symbol: KCNMB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546427.2 | XP_005546484.1 | 3 - 4 | 1080 | 122 | |
| XM_005546428.2 | XP_005546485.1 | 3 - 4 | 888 | 122 | ||
| XM_015445548.1 | XP_015301034.1 | 3 - 4 | 647 | 122 | ||
More Information
Set Membership: |
|

KCNMB3
Genomic Map