Assay Details
| Gene Symbol: TEKT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284892.1 | NP_001271821.1 | 7 - 8 | 959 | 66 | |
| XM_005543977.2 | XP_005544034.1 | 7 - 8 | 1286 | 66 | ||
| XM_005543978.2 | XP_005544035.1 | 7 - 8 | 931 | 66 | ||
| GenBank mRNA | AB169442.1 | - | 7 - 8 | 961 | 66 | |
More Information
Set Membership: |
|

TEKT2
Genomic Map