Assay Details
| Gene Symbol: LOC102115861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441593.1 | XP_015297079.1 | 25 - 26 | 3682 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102115861| Gene Symbol: LOC102115861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441593.1 | XP_015297079.1 | 25 - 26 | 3682 | 126 | |
Set Membership: |
|