Assay Details
Gene Symbol: SPART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585640.2 | XP_005585697.1 | 6 - 7 | 1681 | 79 | |
XM_005585641.2 | XP_005585698.1 | 6 - 7 | 1735 | 79 | ||
XM_005585643.2 | XP_005585700.1 | 6 - 7 | 1654 | 79 | ||
XM_005585644.2 | XP_005585701.1 | 6 - 7 | 2021 | 79 | ||
XM_005585645.2 | XP_005585702.1 | 6 - 7 | 1750 | 79 | ||
XM_015439152.1 | XP_015294638.1 | 7 - 8 | 1769 | 79 | ||
XM_015439153.1 | XP_015294639.1 | 7 - 8 | 1594 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
