Assay Details
Gene Symbol: TMEM131L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556101.2 | XP_005556158.1 | 5 - 6 | 727 | 119 | |
XM_005556102.2 | XP_005556159.1 | 5 - 6 | 493 | 119 | ||
XM_005556103.2 | XP_005556160.1 | 5 - 6 | 509 | 119 | ||
XM_005556104.2 | XP_005556161.1 | 5 - 6 | 485 | 119 | ||
XM_005556105.2 | XP_005556162.1 | 3 - 4 | 353 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
