Assay Details
Gene Symbol: ADNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569349.1 | XP_005569406.1 | 3 - 4 | 393 | 90 | |
XM_015429972.1 | XP_015285458.1 | 3 - 4 | 1625 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
