Assay Details
Gene Symbol: SDF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319586.1 | NP_001306515.1 | 5 - 6 | 1298 | 140 | |
XM_015444118.1 | XP_015299604.1 | 6 - 7 | 1297 | 140 | ||
GenBank mRNA | AB169659.1 | - | 5 - 6 | 1298 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
