Assay Details
| Gene Symbol: CD244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454735.1 | XP_015310221.1 | 6 - 7 | 1122 | 140 | |
| XM_015454739.1 | XP_015310225.1 | 6 - 7 | 1116 | 134 | ||
| XM_015454740.1 | XP_015310226.1 | 6 - 7 | 1107 | 140 | ||
| XM_015454744.1 | XP_015310230.1 | 6 - 7 | 1101 | 134 | ||
| XM_015454750.1 | XP_015310236.1 | 6 - 7 | 1122 | 140 | ||
More Information
Set Membership: |
|

CD244
Genomic Map