Assay Details
| Gene Symbol: HACD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284632.1 | NP_001271561.1 | 2 - 3 | 296 | 104 | |
| GenBank mRNA | AB170635.1 | - | 2 - 3 | 296 | 104 | |
More Information
Set Membership: |
|

HACD3
Genomic Map