Assay Details
| Gene Symbol: CLEC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570100.1 | XP_005570157.1 | 3 - 4 | 523 | 115 | |
| XM_005570101.2 | XP_005570158.1 | 2 - 3 | 424 | 115 | ||
More Information
Set Membership: |
|

CLEC1B
Genomic Map