Assay Details
| Gene Symbol: CHRAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564141.2 | XP_005564198.1 | 1 - 2 | 347 | 97 | |
| XM_015455230.1 | XP_015310716.1 | 1 - 2 | 186 | 97 | ||
More Information
Set Membership: |
|

CHRAC1
Genomic Map