Assay Details
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582681.2 | XP_005582738.2 | 3 - 4 | 1323 | 119 | |
XM_015437546.1 | XP_015293032.1 | 2 - 3 | 1453 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
