Assay Details
Gene Symbol: C19H1orf109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588546.2 | XP_005588603.2 | 2 - 3 | 473 | 116 | |
XM_005588547.2 | XP_005588604.1 | 4 - 5 | 794 | 116 | ||
XM_005588549.2 | XP_005588606.1 | 3 - 4 | 697 | 116 | ||
XM_005588550.2 | XP_005588607.1 | 3 - 4 | 648 | 116 | ||
XM_005588551.2 | XP_005588608.1 | 3 - 4 | 500 | 116 | ||
XM_015440729.1 | XP_015296215.1 | 4 - 5 | 1041 | 116 | ||
XM_015440730.1 | XP_015296216.1 | 5 - 6 | 950 | 116 | ||
XM_015440731.1 | XP_015296217.1 | 4 - 5 | 1032 | 116 | ||
XM_015440732.1 | XP_015296218.1 | 4 - 5 | 1029 | 116 | ||
XM_015440733.1 | XP_015296219.1 | 3 - 4 | 924 | 116 | ||
XM_015440734.1 | XP_015296220.1 | 3 - 4 | 928 | 116 | ||
XM_015440736.1 | XP_015296222.1 | 3 - 4 | 635 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
