Assay Details
| Gene Symbol: PINX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562590.2 | XP_005562647.1 | 5 - 6 | 527 | 141 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PINX1| Gene Symbol: PINX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562590.2 | XP_005562647.1 | 5 - 6 | 527 | 141 | |
Set Membership: |
|