Assay Details
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592254.2 | XP_005592311.1 | 1 - 2 | 90 | 96 | |
XM_005592255.2 | XP_005592312.1 | 1 - 2 | 90 | 96 | ||
XM_005592256.2 | XP_005592313.1 | 1 - 2 | 90 | 96 | ||
XM_005592257.2 | XP_005592314.1 | 1 - 2 | 90 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
