Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005565228.2 | XP_005565285.1 | 2 - 3 | 1869 | 150 | |
XM_005565229.2 | XP_005565286.1 | 2 - 3 | 1655 | 150 | ||
XM_015456071.1 | XP_015311557.1 | 3 - 4 | 1675 | 150 | ||
XM_015456072.1 | XP_015311558.1 | 2 - 3 | 1656 | 150 | ||
XM_015456073.1 | XP_015311559.1 | 2 - 3 | 1656 | 150 | ||
XM_015456074.1 | XP_015311560.1 | 2 - 3 | 1656 | 150 | ||
XM_015456075.1 | XP_015311561.1 | 2 - 3 | 1656 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
