Assay Details
Gene Symbol: POPDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005552255.2 | XP_005552312.1 | 3 - 4 | 996 | 85 | |
XM_015449026.1 | XP_015304512.1 | 4 - 5 | 1136 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
