Assay Details
| Gene Symbol: CHST8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588770.2 | XP_005588827.1 | 2 - 3 | 498 | 118 | |
| XM_005588771.2 | XP_005588828.1 | 2 - 3 | 944 | 118 | ||
More Information
Set Membership: |
|

CHST8
Genomic Map