Assay Details
| Gene Symbol: GRAMD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557645.2 | XP_005557702.1 | 4 - 5 | 551 | 129 | |
| XM_005557646.2 | XP_005557703.1 | 4 - 5 | 551 | 129 | ||
| XM_005557647.2 | XP_005557704.1 | 5 - 6 | 1428 | 129 | ||
| XM_005557648.2 | XP_005557705.1 | 5 - 6 | 1425 | 129 | ||
| XM_005557649.2 | XP_005557706.1 | 4 - 5 | 1381 | 129 | ||
| XM_005557650.2 | XP_005557707.1 | 4 - 5 | 1379 | 129 | ||
| XM_015451837.1 | XP_015307323.1 | 5 - 6 | 594 | 129 | ||
| XM_015451838.1 | XP_015307324.1 | 5 - 6 | 1589 | 129 | ||
More Information
Set Membership: |
|

GRAMD2B
Genomic Map